Silicon(IV) oxide, amorphous, REacton® S.A. 205-245 m²/g, fumed, surface treated S.A. 205-245 m²/g
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous
42782-18
42782-30
42782-A1
CAAA42782-18EA
34.44
CAD
CAAA42782-18
CAAA42782-30
CAAA42782-A1
Silicon(IV) oxide, amorphous, REacton® S.A. 205-245 m²/g, fumed, surface treated S.A. 205-245 m²/g
Silica gel amorphous
Coatings, thermal insulation, silicones
CAS No.: 7631-86-9
Molecular Formula: O2Si
Formula Weight: 60.09
Physical Form: -325 mesh powder
Density: 2.2
Appearance: White
Odor: Odorless
pH: 6
MDL No.: MFCD00011232
Coatings, thermal insulation, silicones
Formula:
SiO₂ MW: 60.08 g/mol Melting Pt: 2230 Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...