Dimethyl sulfoxide, anhydrous ≥99.9%, chemSeal™ for HPLC
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane
Packaged under argon in resealable ChemSeal™ bottles.
MDL: MFCD00002089
Beilstein Registry No.: 506008
Notes: Same HPLC specifications as 22914
Fieser: 1,296 14,150 15,146 16,149 17,127 18,149 20,154 21,180
Purity: 99.9+%
CAS No.: 67-68-5
Molecular Formula: (CH3)2SO
Formula Weight: 78.13
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.101
Refractive Index: 1.4790
Sensitivity: Hygroscopic
Odor: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formula:
(CH₃)₂SO Boiling Pt: 189 °C Melting Pt: 18.4 °C Density: 1.101 Flash Pt: 87 °C (189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...