Copper, gauze woven from wire 20 mesh, Ø 0.41 mm (0.016 in)
Supplier: Thermo Scientific Chemicals
42777-GT
42777-HB
42777-HZ
CAAA42777-GTEA
48.66
CAD
CAAA42777-GT
CAAA42777-HB
CAAA42777-HZ
Copper, gauze woven from wire 20 mesh, Ø 0.41 mm (0.016 in)
Copper
Applications: Filtration, electrodes
CAS No.: 7440-50-8
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00010965
Filtration, electrodes
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...