Cobalt(II) carbonate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42745-18
42745-30
CAAA42745-18EA
138.09
CAD
CAAA42745-18
CAAA42745-30
Cobalt(II) carbonate ≥99.5% (metals basis)
Cobalt (II) carbonate
MDL: MFCD00010940
Purity: 99.5%
CAS No.: 513-79-1
Molecular Formula: CoCO3
Formula Weight: 118.94
Physical Form: -325 Mesh Powder
Density: 4.13
Melting Point: dec.
Merck Reference: 14,2436
UN No.: 3077
MDL No.: MFCD00010940
Formula:
CCoO₃ Melting Pt: Dec. Density: 4.13 Storage Temperature: Ambient |
MDL Number:
MFCD00010940 CAS Number: 513-79-1 EINECS: 208-169-4 UN: 3077 ADR: 9,III Merck Index: 14,2436 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...