Silicon(IV) oxide, amorphous ≥99.8% S.A. 85-115 m²/g, fumed S.A. 85-115 m²/g
Supplier: Thermo Scientific Chemicals
|
Warning
|
42737-18
42737-30
42737-A1
CAAA42737-18EA
48.35
CAD
CAAA42737-18
CAAA42737-30
CAAA42737-A1
Silicon(IV) oxide, amorphous ≥99.8% S.A. 85-115 m²/g, fumed S.A. 85-115 m²/g
Silica gel amorphous
Adhesives, sealants, silicones
CAS No.: 7631-86-9
Molecular Formula: O2Si
Formula Weight: 60.09
Physical Form: -325 Mesh Powder
Density: 2.2
Refractive Index: 1.46
Odor: Odorless
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
Adhesives, sealants, silicones
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...