Potassium hydroxide standardised solution 1.0 N in methanol

Supplier: Thermo Scientific Chemicals
Danger

42736-K2 42736-K7 42736-M4
CAAA42736-K2EA 92.75 CAD
CAAA42736-K2 CAAA42736-K7 CAAA42736-M4
Potassium hydroxide standardised solution 1.0 N in methanol
Potassium hydroxide

MDL: MFCD00003553


CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colorless Odor: Like alcohol UN No.: 3286 MDL No.: MFCD00003553

Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR