Potassium hydroxide standardised solution 0.5 N in methanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
42735-K2
42735-K7
42735-M4
CAAA42735-K2EA
99.54
CAD
CAAA42735-K2
CAAA42735-K7
CAAA42735-M4
Potassium hydroxide standardised solution 0.5 N in methanol
Potassium hydroxide
MDL: MFCD00003553
CAS No.: 1310-58-3
Molecular Formula: KOH
Formula Weight: 56.11
Physical Form: Liquid
Appearance: Colorless
Odor: Like alcohol
UN No.: 3286
MDL No.: MFCD00003553
Formula:
KOH Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...