Silica gel, Pore size: 60 Å 0.062-0.105 mm (150-230 mesh)

Supplier: Thermo Scientific Chemicals
Warning

42727-18 42727-30 42727-A1 42727-A7
CAAA42727-18EA 45.35 CAD
CAAA42727-18 CAAA42727-30 CAAA42727-A1 CAAA42727-A7
Silica gel, Pore size: 60 Å 0.062-0.105 mm (150-230 mesh)
Silica gel

Column chromatography without applied pressure (gravity pulled)


CAS No.: 7631-86-9 Physical Form: -150+230 Mesh, S.A. 500-600m2/g Odor: Odorless MDL No.: MFCD00011232


Column chromatography without applied pressure (gravity pulled)

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR