Silica gel, Pore size: 150 Å, wide pore

Supplier: Thermo Scientific Chemicals
Warning

42726-18 42726-30 42726-A1 42726-A7
CAAA42726-18EA 88.32 CAD
CAAA42726-18 CAAA42726-30 CAAA42726-A1 CAAA42726-A7
Silica gel, Pore size: 150 Å, wide pore
Silica gel

Column chromatography without applied pressure (gravity pulled)


CAS No.: 63231-67-4 Physical Form: -100+200 Mesh, S.A. 350-400m2/g Odor: Odorless MDL No.: MFCD00011232


Column chromatography without applied pressure (gravity pulled)

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR