Silica gel, Pore Size 60 Å 0.075-0.2 mm (70-200 mesh)

Supplier: Thermo Scientific Chemicals
Warning

42723-18 42723-30 42723-A1 42723-A7
CAAA42723-18EA 51.19 CAD
CAAA42723-18 CAAA42723-30 CAAA42723-A1 CAAA42723-A7
Silica gel, Pore Size 60 Å 0.075-0.2 mm (70-200 mesh)
Silica gel

Classic column chromatography with no applied pressure (gravity pulled)


CAS No.: 7631-86-9 Physical Form: -70+200 Mesh Powder, S.A. 470-570m2/g Density: 2.4 Appearance: White Odor: Odorless pH: 6 Melting Point: 1710° Boiling Point: 2230° MDL No.: MFCD00011232


Classic column chromatography with no applied pressure (gravity pulled)

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2,4
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR