Zinc ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

42703-G5 42703-G9
CAAA42703-G5EA 248.06 CAD
CAAA42703-G5 CAAA42703-G9
Zinc ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Zinc

MDL: MFCD00011291


Purity: 99.95% CAS No.: 7440-66-6 Physical Form: ≈0.35g/m Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR