Octyltrimethoxysilane 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42698-06
42698-14
42698-22
CAAA42698-06EA
48.51
CAD
CAAA42698-06
CAAA42698-14
CAAA42698-22
Octyltrimethoxysilane 97+%
Octyltrimethoxysilane
MDL: MFCD00043053
Purity: 97+%
CAS No.: 3069-40-7
Molecular Formula: C11H26O3Si
Formula Weight: 234.41
Flash Point: 102°(216°F)
Physical Form: Liquid
Density: 0.907
Refractive Index: 1.417
Sensitivity: Moisture Sensitive
Appearance: Colorless to yellow
Boiling Point: 75°/0.1mm
MDL No.: MFCD00043053
Formula:
C₁₁H₂₆O₃Si Boiling Pt: 75 °C (0.1 mmHg) Density: 0,907 Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043053 CAS Number: 3069-40-7 EINECS: 221-338-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...