Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Danger

42687-FF 42687-FI
CAAA42687-FFEA 298.62 CAD
CAAA42687-FF CAAA42687-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈0.14g/25x25mm, Packaged in mylar under argon Appearance: Gray to silver Odor: Odorless UN No.: 3178 MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR