Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
42684-FF
42684-FI
CAAA42684-FFEA
1423.58
CAD
CAAA42684-FF
CAAA42684-FI
Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.5 mm (0.02 in)
Palladium Silver alloy (75:25 w%)
MDL: MFCD02091738
Purity: 99.9%
Molecular Formula: Pd:Ag; 75:25 wt%
Physical Form: ≈3.64g/25x25mm
Odor: Odorless
MDL No.: MFCD02091738
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...