Titanium(IV) oxide rutile ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42681-30
42681-A1
42681-A7
CAAA42681-30EA
175.38
CAD
CAAA42681-30
CAAA42681-A1
CAAA42681-A7
Titanium(IV) oxide rutile ≥99.8% (metals basis)
Titanium (IV) oxide rutile
MDL: MFCD00011269
Notes: Tap density 1.0g/cc
Purity: 99.8%
CAS No.: 1317-80-2
Molecular Formula: TiO2, 97% min rutile phase
Formula Weight: 79.90
Physical Form: 0.9-1.6 Micron APS Powder, S.A. 3.3-6.1 m2/g
Density: 4.23
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formula:
O₂Ti Boiling Pt: 2500…3000 °C Melting Pt: 1830…1850 °C Density: 4.23 Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 1317-80-2 EINECS: 215-282-2 Merck Index: 14,9472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...