Platinum iridium alloy (90:10 w%), wire, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
42678-BQ
42678-BU
42678-BY
CAAA42678-BQEA
948
CAD
CAAA42678-BQ
CAAA42678-BU
CAAA42678-BY
Platinum iridium alloy (90:10 w%), wire, Ø 1.0 mm (0.04 in)
Platinum Iridium alloy (90:10 w%)
MDL: MFCD02091683
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.85g/5cm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...