Germanium(IV) isopropoxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42673-03
42673-06
42673-14
CAAA42673-03EA
103.81
CAD
CAAA42673-03
CAAA42673-06
CAAA42673-14
Germanium(IV) isopropoxide ≥99.9% (metals basis)
Germanium (IV) isopropoxide
MDL: MFCD00015339
Purity: 99.9%
CAS No.: 21154-48-3
Molecular Formula: C12H28GeO4
Formula Weight: 308.94
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.025
Refractive Index: 1.414
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Alcohol-like
Boiling Point: 109°/30mm
MDL No.: MFCD00015339
Formula:
C₁₂H₂₈GeO₄ Boiling Pt: 109 °C (30 mmHg) Density: 1,025 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015339 CAS Number: 21154-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...