Cobalt ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

42661-FF 42661-FI
CAAA42661-FFEA 202.24 CAD
CAAA42661-FF CAAA42661-FI
Cobalt ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Cobalt

MDL: MFCD00010935


Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈11.2g/25x25mm Odor: Odorless MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR