Cobalt ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

42658-FF 42658-FI 42658-GH
CAAA42658-FFEA 96.22 CAD
CAAA42658-FF CAAA42658-FI CAAA42658-GH
Cobalt ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Cobalt

MDL: MFCD00010935


Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.56g/25x25mm Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR