Cobalt ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42657-FF
42657-FI
42657-GH
CAAA42657-FFEA
117.24
CAD
CAAA42657-FF
CAAA42657-FI
CAAA42657-GH
Cobalt ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Cobalt
MDL: MFCD00010935
Purity: 99.95%
CAS No.: 7440-48-4
Physical Form: ≈0.28g/25x25mm
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...