1-Octanesulfonic acid sodium salt, hydrate 99+% for HPLC

Supplier: Thermo Scientific Chemicals

42636-06 42636-14 42636-22
CAAA42636-06EA 83.27 CAD
CAAA42636-06 CAAA42636-14 CAAA42636-22
1-Octanesulfonic acid sodium salt, hydrate 99+% for HPLC
1-Octanesulphonic acid sodium salt

Ion-pair reagent for HPLC analysis


Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.


Ion-pair reagent for HPLC analysis

Formula: C₈H₁₇NaO₃S·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4

Order Now

Specification Test Results

Loss on drying 6.9-8.5%
10% water solutionClear Pass Test.
colorless 0.4% EtOH/MeOHClear Pass Test
clear0.4% EtOH/MeOH Clear
IR identification Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR