Titanium ≥99.5% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

42624-22 42624-36 42624-D3
CAAA42624-22EA 148.52 CAD
CAAA42624-22 CAAA42624-36 CAAA42624-D3
Titanium ≥99.5% (metals basis), powder -325 mesh
Titanium

MDL: MFCD00011264


Purity: 99.5% CAS No.: 7440-32-6 Odor: Odorless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR