Copper ≥99.9% (metals basis), spherical powder -100/+325 mesh

Supplier: Thermo Scientific Chemicals
Warning

42623-22 42623-36 42623-A3
CAAA42623-22EA 85.48 CAD
CAAA42623-22 CAAA42623-36 CAAA42623-A3
Copper ≥99.9% (metals basis), spherical powder -100/+325 mesh
Copper

MDL: MFCD00010965 Notes: Oxygen analysis included


Purity: 99.9% CAS No.: 7440-50-8 Appearance: Red Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR