Zirconium(IV) tert-butanolate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate

42583-03 42583-06 42583-14
CAAA42583-03EA 63.52 CAD
CAAA42583-03 CAAA42583-06 CAAA42583-14
Zirconium(IV) tert-butanolate 97+%
Zirconium (IV) tert-butanolate

MDL: MFCD00075085 Fieser: 19,411


Purity: 97+% CAS No.: 2081-12-1 Molecular Formula: C16H36O4Zr Formula Weight: 383.68 Flash Point: 85°(185°F) Physical Form: Liquid Density: 0.985 Sensitivity: Moisture and Light Sensitive Appearance: Yellow Boiling Point: 90°/5mm Beilstein Reference: 3681870 MDL No.: MFCD00075085

Formula: C₁₆H₃₆O₄Zr
MW: 383.68 g/mol
Boiling Pt: 90 °C (5 mmHg)
Density: 0.985
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075085
CAS Number: 2081-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR