Silver(I) oxide 99,99 Metals basis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Silver oxide, Disilver oxide, Silver oxide
42577-03
42577-06
42577-14
42577-49
CAAA42577-03EA
49.3
CAD
CAAA42577-03
CAAA42577-06
CAAA42577-14
CAAA42577-49
Silver(I) oxide 99,99 Metals basis
Silver(I) oxide
MDL: MFCD00003404
Purity: 99.99%
CAS No.: 20667-12-3
Molecular Formula: Ag2O
Formula Weight: 231.74
Physical Form: Powder
Density: 7.2
Appearance: Dark brown
Odor: Odorless
Melting Point: 230° dec.
Merck Reference: 14,8521
UN No.: 1479
MDL No.: MFCD00003404
Formula:
Ag₂O MW: 231.74 g/mol Melting Pt: 230 °C (dec.) Density: 7.2 Storage Temperature: Ambient |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 EINECS: 243-957-1 UN: 1479 ADR: 5.1 II Merck Index: 14,8521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...