Aluminium oxide, α-phase ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
ɑ-Tonerde, α-Alumina
42573-22
42573-36
42573-A3
CAAA42573-22EA
71.42
CAD
CAAA42573-22
CAAA42573-36
CAAA42573-A3
Aluminium oxide, α-phase ≥99.95% (metals basis)
Aluminium oxide, α-phase
MDL: MFCD00003424
Purity: 99.95% min
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 0.25-0.45 Micron APS Powder, S.A. 7.2-8.8m2/g
Refractive Index: 1.765
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...