Aluminium oxide, α-phase ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
ɑ-Tonerde, α-Alumina
42572-22
42572-36
42572-A3
CAAA42572-22EA
70.15
CAD
CAAA42572-22
CAAA42572-36
CAAA42572-A3
Aluminium oxide, α-phase ≥99.9% (metals basis)
Aluminium oxide, α-phase
Suitable for ceramic, electronic ceramic, and ceramic matrix composites
Purity: 99.9%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form:
Suitable for ceramic, electronic ceramic, and ceramic matrix composites
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...