Aluminium oxide, α-phase ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
ɑ-Tonerde, α-Alumina
42571-22
42571-36
42571-A3
CAAA42571-22EA
54.35
CAD
CAAA42571-22
CAAA42571-36
CAAA42571-A3
Aluminium oxide, α-phase ≥99.9% (metals basis)
Aluminium oxide, α-phase
MDL: MFCD00003424
Purity: 99.9%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 20-50 Micron APS, Spherical Powder, S.A. 5-6m2/g
Refractive Index: 1.765
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...