Silica gel, Pore size: 60 Å ≥99.5% (metals basis) 0.040-0.063 mm (230-400 mesh)

Supplier: Thermo Scientific Chemicals
Warning

42570-18 42570-30 42570-A1 42570-A7
CAAA42570-18EA 54.67 CAD
CAAA42570-18 CAAA42570-30 CAAA42570-A1 CAAA42570-A7
Silica gel, Pore size: 60 Å ≥99.5% (metals basis) 0.040-0.063 mm (230-400 mesh)
Silica gel

Column chromatography


Purity: 99.5% CAS No.: 7631-86-9 Physical Form: -230+400 Mesh, S.A. 500-600m2/g Odor: Odorless Merck Reference: 14,8493 MDL No.: MFCD00011232


Column chromatography

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR