Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42558-06 42558-14
CAAA42558-06EA 110.28 CAD
CAAA42558-06 CAAA42558-14
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Zirconium

MDL: MFCD00011303


Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈0.33g/slug Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR