Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
42556-QJ
42556-QK
CAAA42556-QJEA
230.68
CAD
CAAA42556-QJ
CAAA42556-QK
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Zirconium
MDL: MFCD00011303
Purity: 99.95%
CAS No.: 7440-67-7
Physical Form: ≈2.62g/slug
Odor: Odorless
MDL No.: MFCD00011303
Formula:
Zr MW: 91.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...