Digallium trioxide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

42528-06 42528-14 42528-22
CAAA42528-06EA 116.6 CAD
CAAA42528-06 CAAA42528-14 CAAA42528-22
Digallium trioxide ≥99.999% (metals basis), Puratronic®
Digallium trioxide

MDL: MFCD00011020


Purity: 99.999% CAS No.: 12024-21-4 Molecular Formula: Ga2O3 Formula Weight: 187.44 Physical Form: -100 Mesh Powder Density: 6.44 Refractive Index: 1.92 Odor: Odorless Melting Point: 1900° Merck Reference: 14,4346 MDL No.: MFCD00011020

Formula: Ga₂O₃
MW: 187.44 g/mol
Melting Pt: 1900 °C
Density: 6.44
Storage Temperature: Ambient
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7
Merck Index: 14,4346

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR