Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42511-09 42511-18
CAAA42511-09EA 93.85 CAD
CAAA42511-09 CAAA42511-18
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Zirconium

MDL: MFCD00011303


Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈0.16g/slug Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR