Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

42509-14 42509-22
CAAA42509-14EA 88.48 CAD
CAAA42509-14 CAAA42509-22
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Zirconium

MDL: MFCD00011303


Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈1.31g/slug Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR