Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

42508-14 42508-22
CAAA42508-14EA 81.05 CAD
CAAA42508-14 CAAA42508-22
Zirconium ≥99.5% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Zirconium

MDL: MFCD00011303


Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈2.62g/slug Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR