Silver ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
42495-BQ
42495-BU
CAAA42495-BQEA
126.08
CAD
CAAA42495-BQ
CAAA42495-BU
Silver ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Silver
MDL: MFCD00003397
Purity: 99.95%
CAS No.: 7440-22-4
Physical Form: ≈2.06g/cm
Odor: Characteristic
MDL No.: MFCD00003397
Formula:
Ag Boiling Pt: 2212 °C (1013 hPa) Melting Pt: 962 °C Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...