Tantalum ≥99.9% (metals basis), rods, Ø 4.75 mm (0.19 in)
Supplier: Thermo Scientific Chemicals
42494-BQ
42494-BU
CAAA42494-BQEA
199.08
CAD
CAAA42494-BQ
CAAA42494-BU
Tantalum ≥99.9% (metals basis), rods, Ø 4.75 mm (0.19 in)
Tantalum
MDL: MFCD00011252
Purity: 99.9%
CAS No.: 7440-25-7
Physical Form: ≈3.26g/cm
Odor: Odorless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...