Silver 99,998 Metals basis, wire, Premion®, Ø 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

42487-BU 42487-BY 42487-CC
CAAA42487-BUEA 114.08 CAD
CAAA42487-BU CAAA42487-BY CAAA42487-CC
Silver 99,998 Metals basis, wire, Premion®, Ø 1.5 mm (0.06 in)
Silver

MDL: MFCD00003397


Purity: 99.998% CAS No.: 7440-22-4 Physical Form: ≈18.54g/m Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR