Tungsten ≥99.95% (metals basis), powder -200+325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
42477-22
42477-36
42477-A3
CAAA42477-22EA
111.55
CAD
CAAA42477-22
CAAA42477-36
CAAA42477-A3
Tungsten ≥99.95% (metals basis), powder -200+325 mesh
Tungsten
MDL: MFCD00011461
Purity: 99.95%
CAS No.: 7440-33-7
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...