Methoxytrimethylsilane 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethyl(methoxy)silane
42464-09
42464-18
42464-30
CAAA42464-09EA
51.67
CAD
CAAA42464-09
CAAA42464-18
CAAA42464-30
Methoxytrimethylsilane 97+%
Methoxytrimethylsilane
MDL: MFCD00008344
Purity: 97+%
CAS No.: 1825-61-2
Molecular Formula: C4H12OSi
Formula Weight: 104.22
Flash Point: -11°(-12°F)
Physical Form: Liquid
Density: 0.756
Refractive Index: 1.3678
Sensitivity: Moisture Sensitive
Odor: Mild
Boiling Point: 57-58°
UN No.: 1993
MDL No.: MFCD00008344
Formula:
C₄H₁₂OSi MW: 104.22 g/mol Boiling Pt: 57…58 °C Density: 0,756 Flash Pt: –11 °C (–12 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008344 CAS Number: 1825-61-2 EINECS: 217-369-0 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...