Copper ≥99.9% (metals basis), powder -625 mesh (APS 3,25-4,75 µm)

Supplier: Thermo Scientific Chemicals
Warning

42455-18 42455-30
CAAA42455-18EA 132.72 CAD
CAAA42455-18 CAAA42455-30
Copper ≥99.9% (metals basis), powder -625 mesh (APS 3,25-4,75 µm)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: Packaged under argon Appearance: Reddish Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR