N,O-Bis(trimethylsilyl)trifluoroacetamide, anhydrous 99+%, chemSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

Packaged under argon in resealable ChemSeal™ bottles.

42429-06 42429-14 42429-22
CAAA42429-06EA 61.62 CAD
CAAA42429-06 CAAA42429-14 CAAA42429-22
N,O-Bis(trimethylsilyl)trifluoroacetamide, anhydrous 99+%, chemSeal™
N,O-Bis(trimethylsilyl)trifluoroacetamide

MDL: MFCD00008269 Beilstein Registry No.: 3607620


Purity: 99+% CAS No.: 25561-30-2 Molecular Formula: C8H18F3NOSi2 Formula Weight: 257.40 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Physical Form: Liquid Density: 0.97 Refractive Index: 1.3840 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Melting Point: -10° Boiling Point: 145-147° Beilstein Reference: 3607620 UN No.: 2924 MDL No.: MFCD00008269

Formula: C₈H₁₈F₃NOSi₂
MW: 257.4 g/mol
Boiling Pt: 145
Melting Pt: -41
Density: 0.97
Flash Pt: 23 °C (73 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR