(3-Chloropropyl)trimethoxysilane, anhydrous 97+%, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane
Packaged under Argon in resealable ChemSeal™ bottles.
MDL: MFCD00000997
Beilstein Registry No.: 969627
Purity: 97+%
CAS No.: 2530-87-2
Molecular Formula: ClCH2CH2CH2Si(OCH3)3
Formula Weight: 198.72
Flash Point: 78°(172°F)
Physical Form: Liquid
Density: 1.077
Refractive Index: 1.4200
Sensitivity: Moisture Sensitive
Odor: Recognizable
Melting Point: -50°
Boiling Point: 195-197°
Beilstein Reference: 969627
MDL No.: MFCD00000997
Formula:
ClH₂CCH₂CH₂Si(OCH₃)₃ MW: 198.72 g/mol Boiling Pt: 195…197 °C Melting Pt: –50 °C Density: 1.077 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000997 CAS Number: 2530-87-2 EINECS: 219-787-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...