Decamethylcyclopentasiloxane 97%
Supplier: Thermo Scientific Chemicals
Warning
|
42412-18
42412-30
42412-A1
CAAA42412-18EA
46.14
CAD
CAAA42412-18
CAAA42412-30
CAAA42412-A1
Decamethylcyclopentasiloxane 97%
Decamethylcyclopentasiloxane
MDL: MFCD00046966
Notes: Cyclic pentameter-Viscosity: 3.87cSt, dielectric constant: 2.50, Surface tension: 18.0 dynes/cm
Purity: 97%
CAS No.: 541-02-6
Molecular Formula: C10H30O5Si5
Formula Weight: 370.77
Flash Point: 76°(169°F)
Physical Form: Liquid
Density: 0.959
Refractive Index: 1.3982
Appearance: Colorless
Odor: Odorless
Melting Point: -44°
Boiling Point: 210°
Merck Reference: 14,2848
MDL No.: MFCD00046966
Formula:
C₁₀H₃₀O₅Si₅ Boiling Pt: 210 °C Melting Pt: –44 °C Density: 0.959 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046966 CAS Number: 541-02-6 EINECS: 208-764-9 Merck Index: 14,2848 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...