Deuterium chloride 20% (w/w) in D₂O (99.5% D) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

42407-18 42407-18
CAAA42407-18EA 285.98 CAD
CAAA42407-18 AA42407-18
Deuterium chloride 20% (w/w) in D₂O (99.5% D) (w/w)
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
Boiling Pt: 108 °C
Density: 1.20
MDL Number: MFCD00044642
CAS Number: 7698-05-7
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR