Titanium ≥99.995% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals
Warning

42394-06 42394-14
CAAA42394-06EA 112.34 CAD
CAAA42394-06 CAAA42394-14
Titanium ≥99.995% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Titanium

MDL: MFCD00011264


Purity: 99.995% CAS No.: 7440-32-6 Molecular Formula: Ti Physical Form: ≈0.11g/slug Density: 4.5 Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.5
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR