Titanium ≥99.98% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42390-09
42390-18
CAAA42390-09EA
150.89
CAD
CAAA42390-09
CAAA42390-18
Titanium ≥99.98% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Titanium
Evaporation source
Purity: 99.98%
CAS No.: 7440-32-6
Physical Form: ≈0.11g/slug
Odor: Odorless
MDL No.: MFCD00011264
Evaporation source
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...