Iron ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
42385-09
42385-18
CAAA42385-09EA
92.9
CAD
CAAA42385-09
CAAA42385-18
Iron ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Iron
MDL: MFCD00010999
Purity: 99.95%
CAS No.: 7439-89-6
Physical Form: ≈0.2g/slug
MDL No.: MFCD00010999
Formula:
Fe MW: 55.85 g/mol Boiling Pt: 3000 Melting Pt: 1535 Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...