Iron ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

42382-14 42382-22
CAAA42382-14EA 116.45 CAD
CAAA42382-14 CAAA42382-22
Iron ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Iron

MDL: MFCD00010999


Purity: 99.95% CAS No.: 7439-89-6 Physical Form: ≈3.17g/slug MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR