Chloramine-T (sodium salt) trihydrate 98.0-103.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, N-Chloro-4-toluenesulfonamide sodium salt, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
Reagent for selective oxidation of methionine
Purity: 98.0-103.0%
CAS No.: 7080-50-4
Molecular Formula: CH3C6H4SO2N(Cl)Na.3H2O
Formula Weight: 281.69 (227.67anhy)
Flash Point: 192°(377°F)
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odor: Chlorine-like
pH: 8-10
Melting Point: 167-170°
Beilstein Reference: 3924168
Merck Reference: 14,2075
UN No.: 3263
MDL No.: MFCD00149066
Reagent for selective oxidation of methionine
Formula:
CH₃C₆H₄SO₂NClNa·3H₂O Melting Pt: 167…170 °C Density: 1.43 g/cm³ (20 °C) Flash Pt: 192 °C (377 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00149066 CAS Number: 7080-50-4 EINECS: 204-854-7 UN: 3263 ADR: 8,III Merck Index: 14,2075 |
Specification Test Results
pH of a 5% solution | 8.0-10.0 @25° |
Suitable for determination of bromide | Pass test |
Clarity of aqueous solution | Pass test |
Insoluble in alcohol | 0.015 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...